Learn More
Uline 3M 444 Double-Sided Film Tape, Clear
Catalog No. NC2594965
Encompass
Strong bond for plastics, Prepare reports and presentations, Laminate plastic film and foam, attach gaskets to small electronics, Resists temperatures to 180F, Clear polyester film with high tack acrylic adhesive and white paper liner
Catalog No. NC2594965 Supplier Uline Supplier No. S10085
Strong bond for plastics, Prepare reports and presentations, Laminate plastic film and foam, attach gaskets to small electronics, Resists temperatures to 180F, Clear polyester film with high tack acrylic adhesive and white paper liner