Learn More
Electron Microscopy Sciences Double Slot Set Screw Vise Hitachi M4
SDP
Catalog No. 501927123
Encompass_Preferred
Double-slot set screw vise with (2) 1 mm wide x 3 mm deep slots. Clamp thin specimens or cross sections. Effectively holds wafers and thin samples up to 1 mm (.039") thick. The dimensions are 15 mm dia x 6 mm H. Made of aluminum with 2 stainless steel allen set screws. Includes allen wrench.
Type: Hitachi M4
Catalog No. 50-192-7123 Supplier Electron Microscopy Sciences Supplier No. 75943H
Double-slot set screw vise with (2) 1 mm wide x 3 mm deep slots. Clamp thin specimens or cross sections. Effectively holds wafers and thin samples up to 1 mm (.039") thick. The dimensions are 15 mm dia x 6 mm H. Made of aluminum with 2 stainless steel allen set screws. Includes allen wrench.
Type: Hitachi M4