Learn More
Electron Microscopy Sciences Hitachi M4, Hitachi M4
SDP
Catalog No. 501927126
Encompass_Preferred
SEM specimen holder is 32 mm large double slot set screw vise with (2) 2.5 mm wide x 5 mm deep slots. Clamp thin specimens and cross sections. Machined aluminum with 2 stainless steel allen set screws. Includes allen wrench.
Type: Hitachi M4
Size: 32 mm
Catalog No. 50-192-7126 Supplier Electron Microscopy Sciences Supplier No. 75947HH
SEM specimen holder is 32 mm large double slot set screw vise with (2) 2.5 mm wide x 5 mm deep slots. Clamp thin specimens and cross sections. Machined aluminum with 2 stainless steel allen set screws. Includes allen wrench.
Type: Hitachi M4
Size: 32 mm