Learn More
Electron Microscopy Sciences Hitachi M4, Hitachi M4
SDP
Catalog No. 501927125
Encompass_Preferred
Large double-slot set screw vise with (2) 2.5 mm x 5 mm deep slots. Clamp thin specimens and cross sections without conductive paint or tape. Dimensions: 25 x 10 mm (1 x 0.4"). Material: Machined aluminum with 2 stainless steel allen set screws. Includes allen wrench.
Type: Hitachi M4
Catalog No. 50-192-7125 Supplier Electron Microscopy Sciences Supplier No. 75947H
Large double-slot set screw vise with (2) 2.5 mm x 5 mm deep slots. Clamp thin specimens and cross sections without conductive paint or tape. Dimensions: 25 x 10 mm (1 x 0.4"). Material: Machined aluminum with 2 stainless steel allen set screws. Includes allen wrench.
Type: Hitachi M4