Learn More
Electron Microscopy Sciences Razor Blade Double Edge Coated Blade Washed
SDP
Catalog No. NC0991553
Encompass_Preferred
Stainless Steel "PTFE" coated. PERSONNA brand .004" thick, wrapped individually. Washed and degreased reducing foreign material. Clean room classified.
Catalog No. NC0991553 Supplier Electron Microscopy Sciences Supplier No. 72000WA
Stainless Steel "PTFE" coated. PERSONNA brand .004" thick, wrapped individually. Washed and degreased reducing foreign material. Clean room classified.