Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dbm-C5-VC-PAB-MMAE | 1644228-55-6 | 99.5% | 1474.42 | C68H103Br2N11O15 | 10 MG
SDP

Catalog No. 5000201939
Encompass_Preferred

DBM-C5-VC-PAB-MMAE is a drug-linker conjugate used to construct antibody-drug conjugates (ADCs); it combines a C5-VC-PAB cleavable linker with a maleimide DBM attachment and the cytotoxic payload MMAE.

  • Drug-linker conjugate for ADC construction.
  • Purity 99.5%.
  • Molecular weight 1474.42.
  • Chemical formula C68H103Br2N11O15.
  • CAS number 1644228-55-6.
  • Supplied as 10 MG per package.

Catalog No. 50-002-01939 Supplier Medchemexpress LLC Supplier No. HY16408810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

DBM-C5-VC-PAB-MMAE is a drug-linker conjugate used to construct antibody-drug conjugates (ADCs); it combines a C5-VC-PAB cleavable linker with a maleimide DBM attachment and the cytotoxic payload MMAE.

  • Drug-linker conjugate for ADC construction.
  • Purity 99.5%.
  • Molecular weight 1474.42.
  • Chemical formula C68H103Br2N11O15.
  • CAS number 1644228-55-6.
  • Supplied as 10 MG per package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.