Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dbm-mmaf | 1810001-93-4 | 99.7% | 1082.95 g/mol | C49H74Br2N6O11 | 10 MG
SDP

Catalog No. 5000202089
Encompass_Preferred

DBM-MMAF is a drug-linker conjugate that couples the antitubulin payload monomethyl auristatin F (MMAF) to a DBM linker for use in antibody-drug conjugate (ADC) research. It is supplied as a solid with high purity and is intended for preclinical and research applications.

  • Agent-linker conjugate designed for ADC research.
  • Purity: 99.65% as provided by manufacturer.
  • Solid form; soluble in DMSO at 200 mg/mL.
  • Molecular formula C49H74Br2N6O11; molecular weight 1082.95 g/mol.
  • Recommended storage: 4°C, protect from light; in solvent store at -80°C for 6 months or -20°C for 1 month.
  • Available in small milligram amounts suitable for lead generation and conjugation optimization.

Catalog No. 50-002-02089 Supplier Medchemexpress LLC Supplier No. HY13628710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBM-MMAF is a drug-linker conjugate that couples the antitubulin payload monomethyl auristatin F (MMAF) to a DBM linker for use in antibody-drug conjugate (ADC) research. It is supplied as a solid with high purity and is intended for preclinical and research applications.

  • Agent-linker conjugate designed for ADC research.
  • Purity: 99.65% as provided by manufacturer.
  • Solid form; soluble in DMSO at 200 mg/mL.
  • Molecular formula C49H74Br2N6O11; molecular weight 1082.95 g/mol.
  • Recommended storage: 4°C, protect from light; in solvent store at -80°C for 6 months or -20°C for 1 month.
  • Available in small milligram amounts suitable for lead generation and conjugation optimization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.