Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DBM-MMAF | 1810001-93-4 | 99.7% | 1082.95 g/mol | C49H74Br2N6O11 | 50 MG
SDP

Catalog No. 5000202671
Encompass_Preferred

DBM-MMAF is a drug-linker conjugate used to construct antibody-drug conjugates (ADCs). It links the potent antitubulin payload monomethyl auristatin F (MMAF) to a DBM linker, enabling targeted delivery of cytotoxic agents for research and in vitro evaluation.

  • Drug-linker conjugate for ADC synthesis and evaluation.
  • Contains monomethyl auristatin F (MMAF) payload.
  • High purity (99.65%) and chemical formula C49H74Br2N6O11.
  • Soluble in DMSO; store protected from light at 4°C (in solvent: -80°C up to 6 months).
  • Provided as a 50 mg quantity for research use.

Catalog No. 50-002-02671 Supplier Medchemexpress LLC Supplier No. HY13628750MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DBM-MMAF is a drug-linker conjugate used to construct antibody-drug conjugates (ADCs). It links the potent antitubulin payload monomethyl auristatin F (MMAF) to a DBM linker, enabling targeted delivery of cytotoxic agents for research and in vitro evaluation.

  • Drug-linker conjugate for ADC synthesis and evaluation.
  • Contains monomethyl auristatin F (MMAF) payload.
  • High purity (99.65%) and chemical formula C49H74Br2N6O11.
  • Soluble in DMSO; store protected from light at 4°C (in solvent: -80°C up to 6 months).
  • Provided as a 50 mg quantity for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.