Learn More
Lovibond™ DBNPA Test Kit
Catalog No. 09051504 Shop All Tintometer Group Products
Change view
Click to view available options
Volume (Metric):
10 mL
Catalog No. Volume (Metric)
09-051-504 10 mL
1 options
Catalog No. 09-051-504 Supplier Lovibond™ Supplier No. 56K701190
Add to Cart
Add to Cart
Measures DBNPA in water systems from 0 to 6.8 mg/L. Higher ranges can be achieved with dilutions.
  • DBNPA (dibromonitrile propionamide) is a fast-acting biocide that rapidly degrades to non-hazardous by-products, including ammonia, bromide ions, dibromoacetonitrile and dibromoacetic acid. It therefore also meets strict environmental requirements. It is one of the biocides recommended by many guidelines for the treatment of industrial process water.
  • In water, DBNPA kills microorganisms quickly and acts reliably against bacteria, fungi, yeasts, algae and microbiological slime: it can quickly penetrate the microbial cell membrane, act on certain protein genes and thus lead to cell death.
  • The application of the Test Kit is very easy. After pre-filtration of the water sample, a certain number of reagents is added step by step. Further filtration and the addition of reagent tablets follows. The colour change is used to interpret the results with the help of the colour disc in the comparator.
  • Disc Reading: 0.0 / 0.3 / 0.7 / 1.0 / 1.4 / 1.7 / 2.0 / 2.4 / 2.7 / 3.0 / 3.4 / 3.7 / 4.1 / 4.4 / 4.8 / 5.1 / 6.0 / 6.8
Specifications
pH Range 0 to 14
Volume (Metric) 10 mL
Range 0 to 6.8 mg/L DBNPA
Type DBNPA Test