Learn More
Aviva Systems Biology Corp DBP Antibody - N-terminal region (ARP32195_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32195_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DBP. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human DBP. Alternate names: DABPApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Sheep
Catalog No. 50-014-54483