Learn More
Aviva Systems Biology Corp DBP Antibody - C-terminal region (ARP57959_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57959_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DBP. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human DBP. Alternate names: DABPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-286-420