Learn More
Biomatik Corporation Dbp Polyclonal Antibody
Supplier:  Biomatik Corporation CAC11914
Encompass_Preferred
Dbp Polyclonal Antibody is a Human (Homo sapiens) adenovirus C serotype 5 (HAdV-5) (Human (Homo sapiens) adenovirus 5) Polyclonal Antibody to DBP. The Dbp Polyclonal Antibody has been validated for the following applications: ELISA, WB. Host: Rabbit. Species: Human (Homo sapiens) adenovirus C serotype 5 (HAdV-5) (Human (Homo sapiens) adenovirus 5). Immunogen: Recombinant Human Adenovirus C Serotype 5 Dna-Binding Protein (Dbp) , Partial (174-529Aa). Applications: ELISA, WB. Target Synonyms: DBP. Accession Number: P03265. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5783