Learn More
Electron Microscopy Sciences DBP (Dibutyl Phthalate) 450 ML
SDP
Catalog No. 50980357
Encompass_Preferred
CAS #84-74-2 C₆H₄(COOC₄H₉)₂ F.W. 278.35
A plasticizer for epoxy resins.
Catalog No. 50-980-357 Supplier Electron Microscopy Sciences Supplier No. 13110
CAS #84-74-2 C₆H₄(COOC₄H₉)₂ F.W. 278.35
A plasticizer for epoxy resins.