Learn More
Apexbio Technology LLC vitamin D binding protein precrusor (208-218) [Homo sapiens]/[Oryctolagus cuniculus] 25mg
SDP
Catalog No. 5000063054
Encompass_Preferred
Vitamin D binding protein (DBP) precursor (208-218) peptide fragment is derived from the multifunctional serum protein DBP a major plasma protein involved in the systemic transport of vitamin D metabolites DBP facilitates the delivery and bioavailability of vitamin D and its metabolites within tissues thereby influencing calcium homeostasis and bone mineral metabolism Additionally DBP exhibits roles in immunomodulation and innate immune responses The peptide segment from residues 208-218 corresponds specifically to a conserved region known for its functional and binding attributes in humans and rabbit species This peptide fragment serves as a research tool for investigating vitamin D-related physiological pathways calcium metabolism mechanisms and immune function interactions in biomedical studies
Catalog No. 50-000-63054 Supplier Apexbio Technology LLC Supplier No. A109825
Vitamin D binding protein (DBP) precursor (208-218) peptide fragment is derived from the multifunctional serum protein DBP a major plasma protein involved in the systemic transport of vitamin D metabolites DBP facilitates the delivery and bioavailability of vitamin D and its metabolites within tissues thereby influencing calcium homeostasis and bone mineral metabolism Additionally DBP exhibits roles in immunomodulation and innate immune responses The peptide segment from residues 208-218 corresponds specifically to a conserved region known for its functional and binding attributes in humans and rabbit species This peptide fragment serves as a research tool for investigating vitamin D-related physiological pathways calcium metabolism mechanisms and immune function interactions in biomedical studies