Learn More
ABclonal Technology DBP Rabbit pAb
SDP
Supplier:  ABclonal Technology A15270500UL
Encompass_Preferred
The protein encoded by this gene is a member of the PAR bZIP transcription factor family and binds to specific sequences in the promoters of several genes, such as albumin, CYP2A4, and CYP2A5. The encoded protein can bind DNA as a homo- or heterodimer and is involved in the regulation of some circadian rhythm genes.
Catalog No. 50-001-06612