Learn More
Echelon Biosciences Research Labs DBPC (PLA2 Substrate) 100 ug
SDP
Supplier:  Echelon Biosciences Research Labs L3000100UG
Encompass
The PLA2 substrate DBPC, is a sensitive probe for a continuous fluorogenic detection applicable to both in vitro and cell-based in situ screening assays.Phospholipase A2 (PLA2) is the enzyme which cleaves phospholipids to produce lysophospholipids and free fatty acids. The PLA2 family of enzymes is known to be at least 10 distinct members. sPLA2-V and sPLA2-X are selectively expressed in human airway epithelium and sPLA2-X in various immune cells. The basal expression of a third enzyme, sPLA2-IIA is low but becomes highly expressed during inflammation and sepsis. This enzyme has become associated with allergic rhinitis, rheumatoid arthritis, septic shock and ARDS. PLA2-IIA represents a target for the treatment of inflammatory disease. This item is non returnable
Catalog No. 50-720-504