Learn More
TARGETMOL CHEMICALS INC DBPR112 5MG
SDP
Catalog No. 502581610
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DBPR112 is an orally active furanopyrimidine-based EGFR inhibitor with IC50s of 15 nM and 48 nM for EGFRWT and EGFRL858R/T790M respectively. DBPR112 can occupy the ATP-binding site. DBPR112 has significant antitumor efficacy. purity: 98%
Catalog No. 50-258-1610 Supplier TARGETMOL CHEMICALS INC Supplier No. T109655MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. DBPR112 is an orally active furanopyrimidine-based EGFR inhibitor with IC50s of 15 nM and 48 nM for EGFRWT and EGFRL858R/T790M respectively. DBPR112 can occupy the ATP-binding site. DBPR112 has significant antitumor efficacy. purity: 98%