Learn More
Medchemexpress LLC HY-128778 5mg Medchemexpress, DBPR112 CAS:1226549-49-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1287785MG
Encompass_Preferred
Medchemexpress, HY-128778 5mg DBPR112 CAS:1226549-49-0 DBPR112 is an orally active furanopyrimidine-based EGFR inhibitor with IC50s of 15 nM and 48 nM for EGFRWT and EGFRL858R/T790M, respectively. DBPR112 can occupy the ATP-binding site. DBPR112 has significant antitumor efficacy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1183