Learn More
Selleck Chemical LLC DC-05 S3543-25mg
SDP
Catalog No. 503108777
Encompass_Preferred
DC-05 is a DNA methyltransferase 1 (DNMT1) inhibitor with an IC50 and a Kd of 10 3 M and 1 09 M respectively
Catalog No. 50-310-8777 Supplier Selleck Chemical LLC Supplier No. S354325MG
DC-05 is a DNA methyltransferase 1 (DNMT1) inhibitor with an IC50 and a Kd of 10 3 M and 1 09 M respectively