Learn More
Medchemexpress LLC HY-12746 5mg Medchemexpress, DC-05 CAS:890643-16-0 Purity:>98%
SDP
Catalog No. 502081130
Encompass_Preferred
Medchemexpress, HY-12746 5mg DC-05 CAS:890643-16-0 DC-05 is a DNA methyltransferase 1 (DNMT1) inhibitor, with an IC50 and a Kd of 10.3 μM and 1.09 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1130 Supplier Medchemexpress LLC Supplier No. HY127465MG
Medchemexpress, HY-12746 5mg DC-05 CAS:890643-16-0 DC-05 is a DNA methyltransferase 1 (DNMT1) inhibitor, with an IC50 and a Kd of 10.3 μM and 1.09 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.