Learn More
Krayden DC 184 KIT 3.9KG/8.8LB
Catalog No. NC9561957
Encompass_Preferred
SYLGARD(R) 184 SILICONE ELASTOMER BASE ,8.8 lb, Dow Corning 184 8.8 lb,Two-part; 10:1 mix; minimal shrinkage; no exothermduring cure; no solvents or cure by-products; deep section cure; repairable; good dielectric properties
Catalog No. NC9561957 Supplier Krayden Supplier No. DC2065622
SYLGARD(R) 184 SILICONE ELASTOMER BASE ,8.8 lb, Dow Corning 184 8.8 lb,Two-part; 10:1 mix; minimal shrinkage; no exothermduring cure; no solvents or cure by-products; deep section cure; repairable; good dielectric properties