Learn More
Promocell GmbH PromoCell DC Base Medium XF 250 ml, with C-39854 attached to flask
Catalog No. 50306293
Encompass
The PromoCell Dendritic Cell (DC) Generation Media has been developed for the easy and efficient generation of immature as well as fully mature myeloid Dendritic Cells from peripheral blood monocytes. Both the DC Base Medium XF and DC Generation Medium XF (Ready-to-use) feature a serum-free and xeno-free formulation for use with freshly isolated cellsThe DC Base Medium XF consist of a bottle of Basal Medium and one vial of SupplementMix. However, it does not include cytokines and must therefore be supplemented according to the user's individual needs.The DC Generation Medium XF (Ready-to-use) consists of a bottle of Basal Medium, one vial of SupplementMix, and CytokineMix containing all necessary differentiation factors.
Catalog No. 50-306-293 Supplier Promocell GmbH Supplier No. C28054
The PromoCell Dendritic Cell (DC) Generation Media has been developed for the easy and efficient generation of immature as well as fully mature myeloid Dendritic Cells from peripheral blood monocytes. Both the DC Base Medium XF and DC Generation Medium XF (Ready-to-use) feature a serum-free and xeno-free formulation for use with freshly isolated cellsThe DC Base Medium XF consist of a bottle of Basal Medium and one vial of SupplementMix. However, it does not include cytokines and must therefore be supplemented according to the user's individual needs.The DC Generation Medium XF (Ready-to-use) consists of a bottle of Basal Medium, one vial of SupplementMix, and CytokineMix containing all necessary differentiation factors.