Learn More
Medchemexpress LLC DC-BPi-03 | 2758411-46-8 | C14H14N4O2S | 10 MM/1 ML
SDP
Catalog No. 5000374200
Encompass_Preferred
DC-BPi-03 is a potent BPTF-BRD inhibitor with an IC50 of 698.3 nM and a Kd of 2.81 μM. It is for research use only and not sold to patients.
  • Potent BPTF-BRD inhibitor
  • IC50 of 698.3 nM
  • Kd of 2.81 μM
  • Solid appearance
  • White to off-white color
  • Purity of 99.77%
  • For research use only
Catalog No. 50-003-74200 Supplier Medchemexpress LLC Supplier No. HY15021110MM/1ML
DC-BPi-03 is a potent BPTF-BRD inhibitor with an IC50 of 698.3 nM and a Kd of 2.81 μM. It is for research use only and not sold to patients.
  • Potent BPTF-BRD inhibitor
  • IC50 of 698.3 nM
  • Kd of 2.81 μM
  • Solid appearance
  • White to off-white color
  • Purity of 99.77%
  • For research use only