Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DC-BPi-11 hydrochloride | 00-00-0 | 97.0% | 433.95 g·mol⁻¹ | C20H24ClN5O2S | 5 MG
SDP

Catalog No. 5000220975
Encompass_Preferred

DC-BPi-11 hydrochloride is a small-molecule BPTF inhibitor supplied as a hydrochloride salt in powder form for research use. It has demonstrated inhibition of leukemia cell proliferation and a reported IC50 of approximately 698 nM, with handling and storage guidance provided for both solid and solution states.

  • Inhibitor of bromodomain PHD finger transcription factor (BPTF).
  • Reported IC50 ≈ 698 nM.
  • Powder hydrochloride salt form.
  • Molecular formula C20H24ClN5O2S.
  • Molecular weight 433.95 g·mol⁻¹.
  • Purity 97.0%.
  • Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Available as a 5 mg sample pack.

Catalog No. 50-002-20975 Supplier Medchemexpress LLC Supplier No. HY141703A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DC-BPi-11 hydrochloride is a small-molecule BPTF inhibitor supplied as a hydrochloride salt in powder form for research use. It has demonstrated inhibition of leukemia cell proliferation and a reported IC50 of approximately 698 nM, with handling and storage guidance provided for both solid and solution states.

  • Inhibitor of bromodomain PHD finger transcription factor (BPTF).
  • Reported IC50 ≈ 698 nM.
  • Powder hydrochloride salt form.
  • Molecular formula C20H24ClN5O2S.
  • Molecular weight 433.95 g·mol⁻¹.
  • Purity 97.0%.
  • Storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Available as a 5 mg sample pack.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.