Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DC-BPi-11 hydrochloride | 00-00-0 | 97.0% | C20H24ClN5O2S | 10MG
SDP

Catalog No. 5000261227
Encompass_Preferred

DC-BPi-11 hydrochloride is a research-grade small-molecule inhibitor of the bromodomain PHD finger transcription factor (BPTF) used in epigenetics and cancer research. It has demonstrated inhibition of leukemia cell proliferation and a reported IC50 of 698 nM against BPTF. The compound is supplied as a powder and analytical batch documentation reports a purity of 97.0%.

  • Inhibits BPTF with an IC50 of 698 nM.
  • Demonstrates inhibition of leukemia cell proliferation.
  • Supplied as a powder for handling and storage flexibility.
  • Powder stable at -20°C for up to 3 years.
  • High reported purity: 97.0%.
  • Chemical formula C20H24ClN5O2S; molecular weight 433.95.

Catalog No. 50-002-61227 Supplier Medchemexpress LLC Supplier No. HY141703A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DC-BPi-11 hydrochloride is a research-grade small-molecule inhibitor of the bromodomain PHD finger transcription factor (BPTF) used in epigenetics and cancer research. It has demonstrated inhibition of leukemia cell proliferation and a reported IC50 of 698 nM against BPTF. The compound is supplied as a powder and analytical batch documentation reports a purity of 97.0%.

  • Inhibits BPTF with an IC50 of 698 nM.
  • Demonstrates inhibition of leukemia cell proliferation.
  • Supplied as a powder for handling and storage flexibility.
  • Powder stable at -20°C for up to 3 years.
  • High reported purity: 97.0%.
  • Chemical formula C20H24ClN5O2S; molecular weight 433.95.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.