Learn More
Medchemexpress LLC DC-BPi-11 (hydrochloride) | 2758411-61-7 | 97.0% | C20H24ClN5O2S | 25 MG
SDP
Catalog No. 5000352804
Encompass_Preferred
DC-BPi-11 hydrochloride is an inhibitor of bromodomain PHD finger transcription factor (BPTF) with an IC50 value of 698 nM. It shows remarkable inhibition against leukemia cell proliferation.
  • Inhibitor of bromodomain PHD finger transcription factor (BPTF)
  • Shows remarkable inhibition against leukemia cell proliferation
  • Decreases downstream oncogene expression
  • Dose-dependently decreases c-Myc protein level
  • Induces cell apoptosis by arresting the cell cycle at G1 phase
  • Minimal effects on normal cells
Catalog No. 50-003-52804 Supplier Medchemexpress LLC Supplier No. 25MGHY141703A
DC-BPi-11 hydrochloride is an inhibitor of bromodomain PHD finger transcription factor (BPTF) with an IC50 value of 698 nM. It shows remarkable inhibition against leukemia cell proliferation.
  • Inhibitor of bromodomain PHD finger transcription factor (BPTF)
  • Shows remarkable inhibition against leukemia cell proliferation
  • Decreases downstream oncogene expression
  • Dose-dependently decreases c-Myc protein level
  • Induces cell apoptosis by arresting the cell cycle at G1 phase
  • Minimal effects on normal cells