Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DC-BPi-11 hydrochloride | 97.0% | C20H24ClN5O2S | 1 ML
SDP

Catalog No. 5000356962
Encompass_Preferred

DC-BPi-11 hydrochloride is an inhibitor of bromodomain PHD finger transcription factor (BPTF) with an IC50 of 698 nM. It significantly inhibits leukemia cell proliferation in human leukemia MV-4-11 cells (EC50 120 nM, IC50 0.89 μM). The compound decreases downstream oncogene expression and c-Myc protein levels, with minimal effects on normal cells. Intended for research use only.

  • Inhibits bromodomain PHD finger transcription factor (BPTF)
  • IC50 of 698 nM against BPTF
  • Inhibits leukemia cell proliferation
  • Decreases oncogene expression and c-Myc protein
  • Minimal effects on normal cells
  • Off-white to light yellow solid appearance
  • Molecular weight: 433.95
  • Chemical formula: C20H24ClN5O2S

Catalog No. 50-003-56962 Supplier Medchemexpress LLC Supplier No. HY141703A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DC-BPi-11 hydrochloride is an inhibitor of bromodomain PHD finger transcription factor (BPTF) with an IC50 of 698 nM. It significantly inhibits leukemia cell proliferation in human leukemia MV-4-11 cells (EC50 120 nM, IC50 0.89 μM). The compound decreases downstream oncogene expression and c-Myc protein levels, with minimal effects on normal cells. Intended for research use only.

  • Inhibits bromodomain PHD finger transcription factor (BPTF)
  • IC50 of 698 nM against BPTF
  • Inhibits leukemia cell proliferation
  • Decreases oncogene expression and c-Myc protein
  • Minimal effects on normal cells
  • Off-white to light yellow solid appearance
  • Molecular weight: 433.95
  • Chemical formula: C20H24ClN5O2S

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.