Learn More
AdipoGen (-)-(8,8-Dichlorocamphorylsulfonyl) oxaziridine, 139628-16-3, MFCD17214220, 25 g
SDP
Supplier:  AdipoGen CDXD0021G025
Encompass_Preferred
MF: C10H13Cl2NO3S, MW: 298.19, ≥95%, Long Term Storage: -20°C. Chiral oxidizing agent. Used for asymmetric synthesis as chiral catalyst. Hydroxylation reagent.
Catalog No. 50-117-7357