Learn More
Medchemexpress LLC DC-Chol hydrochloride | 166023-21-8 | >99.9% | 537.26 | 250 MG
SDP
Catalog No. 5000352285
Encompass_Preferred
DC-Chol hydrochloride is a cationic lipid that inhibits Aβ40 fibril formation and amyloidogenesis of oxidized hCT. It enhances the body's immune response to antigens by inducing the production of Th1 (IL-2 and IFN-γ) and Th2 (IL-5) cytokines. This compound is also utilized as a gene delivery vector.
  • Can inhibit Aβ40 fibril formation
  • Strongly inhibits amyloidogenesis of oxidized hCT in a dose-dependent manner
  • Induces production of Th1 (IL-2 and IFN-γ) and Th2 (IL-5) cytokines
  • Enhances the body's immune response to antigens
  • Used as a gene delivery vector
  • Can be used in research for hepatitis B vaccines to improve vaccine immunity
Catalog No. 50-003-52285 Supplier Medchemexpress LLC Supplier No. 250MGHY137131
DC-Chol hydrochloride is a cationic lipid that inhibits Aβ40 fibril formation and amyloidogenesis of oxidized hCT. It enhances the body's immune response to antigens by inducing the production of Th1 (IL-2 and IFN-γ) and Th2 (IL-5) cytokines. This compound is also utilized as a gene delivery vector.
  • Can inhibit Aβ40 fibril formation
  • Strongly inhibits amyloidogenesis of oxidized hCT in a dose-dependent manner
  • Induces production of Th1 (IL-2 and IFN-γ) and Th2 (IL-5) cytokines
  • Enhances the body's immune response to antigens
  • Used as a gene delivery vector
  • Can be used in research for hepatitis B vaccines to improve vaccine immunity