Learn More
Medchemexpress LLC Dc-chol hydrochloride | 166023-21-8 | 99.9% | 537.26 | 50 MG
SDP
Catalog No. 5000348722
Encompass_Preferred
DC-Chol hydrochloride is a cationic lipid that can inhibit Aβ40 fibril formation and amyloidogenesis. It enhances the body's immune response by inducing Th1 (IL-2 and IFN-γ) and Th2 (IL-5) cytokines, making it suitable for various research applications.
  • Inhibits Aβ40 fibril formation and amyloidogenesis.
  • Induces Th1 and Th2 cytokine production to enhance immune response.
  • Used as a gene delivery vector.
  • Utilized in research for hepatitis B vaccines to improve vaccine immunity.
Catalog No. 50-003-48722 Supplier Medchemexpress LLC Supplier No. HY13713150MG
DC-Chol hydrochloride is a cationic lipid that can inhibit Aβ40 fibril formation and amyloidogenesis. It enhances the body's immune response by inducing Th1 (IL-2 and IFN-γ) and Th2 (IL-5) cytokines, making it suitable for various research applications.
  • Inhibits Aβ40 fibril formation and amyloidogenesis.
  • Induces Th1 and Th2 cytokine production to enhance immune response.
  • Used as a gene delivery vector.
  • Utilized in research for hepatitis B vaccines to improve vaccine immunity.