Learn More
Cayman Chemical DC-Chol (hydrochloride), Purity: ≥95%, CAS: 166023-21-8, 100 mg
SDP
Catalog No. 502965378
Encompass_Preferred
A cationic cholesterol derivative; as a component of lipoplexes with 1,2-DOPE and mRNA, has been used for transfection of A549 cells without affecting cell viability; incubation of liposomes or lipoplexes with human whole blood has no significant effect on neutrophil elastase or β-thromboglobulin levels or the number of platelets and red and white blood cells, indicating hemocompatibility; has been widely used in the synthesis of liposomes for the delivery of siRNA, DNA, and chemotherapeutic agents into cells and mice
Catalog No. 50-296-5378 Supplier Cayman Chemical Supplier No. 16943100 MG
A cationic cholesterol derivative; as a component of lipoplexes with 1,2-DOPE and mRNA, has been used for transfection of A549 cells without affecting cell viability; incubation of liposomes or lipoplexes with human whole blood has no significant effect on neutrophil elastase or β-thromboglobulin levels or the number of platelets and red and white blood cells, indicating hemocompatibility; has been widely used in the synthesis of liposomes for the delivery of siRNA, DNA, and chemotherapeutic agents into cells and mice