Learn More
Grainger DC Hour Meter LCD Round Resettable
Catalog No. 502767691
Encompass_Preferred
The ENM Hour Meter is an electromechanical device that records the operating hours of equipment. It has a 6-digit LCD display with large 0.5" digits for easy viewing. The meter operates on 8-30V DC power and can record up to 99,999.9 hours, after which it rolls over to 0. It has an accuracy of ±0.02% and a pulse count input. The compact 2.36" x 1.57" x 1.06" plastic case is dustproof and water resistant. Ideal for tracking maintenance schedules on machinery, equipment, and other assets.
Catalog No. 50-276-7691 Supplier Grainger Supplier No. 16T957
The ENM Hour Meter is an electromechanical device that records the operating hours of equipment. It has a 6-digit LCD display with large 0.5" digits for easy viewing. The meter operates on 8-30V DC power and can record up to 99,999.9 hours, after which it rolls over to 0. It has an accuracy of ±0.02% and a pulse count input. The compact 2.36" x 1.57" x 1.06" plastic case is dustproof and water resistant. Ideal for tracking maintenance schedules on machinery, equipment, and other assets.