Learn More
Cayman Chemical DC-LC3in-D5, Purity: ≥95%, CAS: 2868312-73-4, 10 mg
SDP
Catalog No. 502919011
Encompass_Preferred
An inhibitor of LC3 (IC50 = 200 nM); selectively binds to LC3-I and LC3-II over GABARAP, SCARB2, and PON2 in an ABPP assay using HeLa cells; inhibits LC3-II lipidation, autophagosome formation, and the protein-protein interaction between LC3-II and ATG7 in HeLa cells at 25 µM
Catalog No. 50-291-9011 Supplier Cayman Chemical Supplier No. 3833910 MG
An inhibitor of LC3 (IC50 = 200 nM); selectively binds to LC3-I and LC3-II over GABARAP, SCARB2, and PON2 in an ABPP assay using HeLa cells; inhibits LC3-II lipidation, autophagosome formation, and the protein-protein interaction between LC3-II and ATG7 in HeLa cells at 25 µM