Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dc-lc3in-d5 | 2868312-73-4 | 98.6% | 397.30 | C19H22Cl2N2O3 | 5 MG
SDP

Catalog No. 5000221760
Encompass_Preferred

DC-LC3in-D5 is a small-molecule autophagy inhibitor that attenuates LC3B lipidation by binding LC3B and disrupting LC3B-LBP2 interactions (reported IC50 ≈ 200 nM). It is supplied as a solid for research use and is used in cellular and biochemical studies of autophagy, antiviral research, and cancer combination studies.

  • Inhibits LC3B lipidation (IC50 ≈ 200 nM).
  • High purity (98.62%).
  • Molecular formula C19H22Cl2N2O3; molecular weight 397.30.
  • Soluble in DMSO (35.71 mg/mL) with ultrasonic warming; heat to 60°C may be used.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).

Catalog No. 50-002-21760 Supplier Medchemexpress LLC Supplier No. HY1418825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DC-LC3in-D5 is a small-molecule autophagy inhibitor that attenuates LC3B lipidation by binding LC3B and disrupting LC3B-LBP2 interactions (reported IC50 ≈ 200 nM). It is supplied as a solid for research use and is used in cellular and biochemical studies of autophagy, antiviral research, and cancer combination studies.

  • Inhibits LC3B lipidation (IC50 ≈ 200 nM).
  • High purity (98.62%).
  • Molecular formula C19H22Cl2N2O3; molecular weight 397.30.
  • Soluble in DMSO (35.71 mg/mL) with ultrasonic warming; heat to 60°C may be used.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.