Learn More
Medchemexpress LLC DC-LC3in-D5 | 2868312-73-4 | 98.6% | C19H22Cl2N2O3 | 10 MM 1 ML
SDP
Catalog No. 5000353237
Encompass_Preferred
DC-LC3in-D5 is an autophagy inhibitor that functions by attenuating LC3B lipidation. It binds with LC3B and disrupts the LC3B-LBP2 interaction with an IC50 of 200 nM. This compound may be useful in anti-HCV or combination cancer research through its autophagy-inhibiting properties.
  • Attenuates LC3B lipidation
  • Disrupts LC3B-LBP2 interaction
  • Demonstrates high selectivity to LC3A/B in proteome
  • Exhibits potent covalent reactivity and selectivity to LC3A/B in HeLa cells
  • Inhibits autophagic vesicle formation
  • Leads to accumulation of p62
Catalog No. 50-003-53237 Supplier Medchemexpress LLC Supplier No. HY14188210MM/1ML
DC-LC3in-D5 is an autophagy inhibitor that functions by attenuating LC3B lipidation. It binds with LC3B and disrupts the LC3B-LBP2 interaction with an IC50 of 200 nM. This compound may be useful in anti-HCV or combination cancer research through its autophagy-inhibiting properties.
  • Attenuates LC3B lipidation
  • Disrupts LC3B-LBP2 interaction
  • Demonstrates high selectivity to LC3A/B in proteome
  • Exhibits potent covalent reactivity and selectivity to LC3A/B in HeLa cells
  • Inhibits autophagic vesicle formation
  • Leads to accumulation of p62