Learn More
Medchemexpress LLC DC-LC3in-D5 | 2868312-73-4 | 98.6% | C19H22Cl2N2O3 | 100 MG
SDP
Catalog No. 5000355357
Encompass_Preferred
DC-LC3in-D5 is an autophagy inhibitor that attenuates LC3B lipidation by binding with LC3B and disrupting the LC3B-LBP2 interaction with an IC50 of 200 nM. This compound demonstrates high selectivity to LC3A/B in the proteome and exhibits potent covalent reactivity and selectivity to LC3A/B in HeLa cells. It may contribute to anti-HCV or combination research in cancer through inhibiting autophagy.
  • Acts as an autophagy inhibitor by attenuating LC3B lipidation
  • Binds with LC3B and disrupts the LC3B-LBP2 interaction
  • Demonstrates high selectivity to LC3A/B in the proteome
  • Exhibits potent covalent reactivity and selectivity to LC3A/B in HeLa cells
  • Treatment of HeLa cells with 3-30 μM results in disruption of LC3B lipidation, inhibition of autophagic vesicle formation, and accumulation of p62
Catalog No. 50-003-55357 Supplier Medchemexpress LLC Supplier No. HY141882100MG
DC-LC3in-D5 is an autophagy inhibitor that attenuates LC3B lipidation by binding with LC3B and disrupting the LC3B-LBP2 interaction with an IC50 of 200 nM. This compound demonstrates high selectivity to LC3A/B in the proteome and exhibits potent covalent reactivity and selectivity to LC3A/B in HeLa cells. It may contribute to anti-HCV or combination research in cancer through inhibiting autophagy.
  • Acts as an autophagy inhibitor by attenuating LC3B lipidation
  • Binds with LC3B and disrupts the LC3B-LBP2 interaction
  • Demonstrates high selectivity to LC3A/B in the proteome
  • Exhibits potent covalent reactivity and selectivity to LC3A/B in HeLa cells
  • Treatment of HeLa cells with 3-30 μM results in disruption of LC3B lipidation, inhibition of autophagic vesicle formation, and accumulation of p62