Learn More
Medchemexpress LLC Dc-lc3in-d5 | 2868312-73-4 | 98.6% | C19H22Cl2N2O3 | 10MG
SDP
Catalog No. 5000258494
Encompass_Preferred
DC-LC3in-D5 is a deuterated research compound that acts as an autophagy inhibitor by attenuating LC3B lipidation and disrupting LC3B interactions. It is provided as a solid for laboratory research and is characterized by high purity and defined storage recommendations.
Catalog No. 50-002-58494 Supplier Medchemexpress LLC Supplier No. HY14188210MG
DC-LC3in-D5 is a deuterated research compound that acts as an autophagy inhibitor by attenuating LC3B lipidation and disrupting LC3B interactions. It is provided as a solid for laboratory research and is characterized by high purity and defined storage recommendations.