Learn More
Sigma Aldrich Fine Chemicals Biosciences DC Mass Spect GoldR Serum
Catalog No. 502904463
Encompass
DC Mass Spect Gold is processed from human plasma through a validated process that reduces the Hormone and Steroid levels please see values below. This validated process maintains the inherent characteristics of the human plasma resulting in a pool that is consistent with patient samples. The human plasma is collected from donors at fDA licensed facilities. Each donor is screened for donor eligibility according to fDA regulations. Each unit is tested and found negative for HIV1/2-ab HBsAg HCV-ab HIV-1 (NAT) HCV (NAT) and RPR. The plasmais inspected for conformance to raw material specifications. The plasma is processed according to a batch record. The finished product is analyzed and inspected according to quality control procedures for conformance to specifications. Records from donor unit(s) to final product are maintained according to established procedures. These records include donor records raw material inspection records batch records testing records (baseline through fi
Catalog No. 50-290-4463 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MSG4000100ML
DC Mass Spect Gold is processed from human plasma through a validated process that reduces the Hormone and Steroid levels please see values below. This validated process maintains the inherent characteristics of the human plasma resulting in a pool that is consistent with patient samples. The human plasma is collected from donors at fDA licensed facilities. Each donor is screened for donor eligibility according to fDA regulations. Each unit is tested and found negative for HIV1/2-ab HBsAg HCV-ab HIV-1 (NAT) HCV (NAT) and RPR. The plasmais inspected for conformance to raw material specifications. The plasma is processed according to a batch record. The finished product is analyzed and inspected according to quality control procedures for conformance to specifications. Records from donor unit(s) to final product are maintained according to established procedures. These records include donor records raw material inspection records batch records testing records (baseline through fi