Learn More
Sterlitech Corporation Direct Contact Membrane Distillation Cell, Acrylic
SDP
Catalog No. 501952350
Encompass_Preferred
Sterlitech’s “plug and play” DCMD test cell is a bench-scale cross/tangential flow test cell that mimics the conditions representative of a large-scale DCMD system while using minimal amounts of membrane or product.
Catalog No. 50-195-2350 Supplier Sterlitech Corporation Supplier No. 1160007
Sterlitech’s “plug and play” DCMD test cell is a bench-scale cross/tangential flow test cell that mimics the conditions representative of a large-scale DCMD system while using minimal amounts of membrane or product.