Learn More
GSC Go Science Crazy DC Motor with Leads
SCI_ED
Catalog No. S24556 Shop All GSC Products
Click to view available options
Voltage:
4.5 to 6.0 DC
Catalog No. Voltage
S24556 4.5 to 6.0 DC
1 options
This item is not returnable. View return policy
Catalog No. S24556 Supplier GSC Go Science Crazy Supplier No. 428010
This item is not returnable. View return policy
Choose the GSC DC Motor with Leads for providing a compact powersource for classroom experiments. This little, yet powerful, motor is ideal for hundreds of electrical experiments and projects.
  • Housing: 2.38mcm D x 3.05cm L
  • Overall length: 4.5cm including the .2cm shaft
  • Voltage requirements: 4.5 to 6.0 DC
  • 0mm long red and black connector cord with insulator
Specifications
For Use With (Application) Classroom experiments
Voltage 4.5 to 6.0 DC
Product Type Motor
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov