Learn More
Bio Rad Laboratories DC, PROTEIN ASSAY KIT. LIQUID
Catalog No. NC9372425
Encompass
Bio-Rad DC Protein Assay Reagents Package, Contains 250mL alkaline copper tartrate solution, 2L dilute Folin reagent, 5mL surfactant solution
Catalog No. NC9372425 Supplier Bio Rad Laboratories Supplier No. 5000116
Bio-Rad DC Protein Assay Reagents Package, Contains 250mL alkaline copper tartrate solution, 2L dilute Folin reagent, 5mL surfactant solution