Learn More
Bio Rad Laboratories DC PROTEIN ASSAY REAGENT A
Catalog No. NC9746741
Encompass
The protein assay reagent A, alkaline copper tartrate solution, is used in the DC protein assay, which quantitates proteins following detergent solubilization. Supplied as 250 ml alkaline copper tartrate. (500-0113)
Catalog No. NC9746741 Supplier Bio Rad Laboratories Supplier No. 5000113
The protein assay reagent A, alkaline copper tartrate solution, is used in the DC protein assay, which quantitates proteins following detergent solubilization. Supplied as 250 ml alkaline copper tartrate. (500-0113)