Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Rad Laboratories DC PROTEIN ASSAY-REAGENT B 1L

Catalog No. NC9854618
Encompass

Protein Assay Reagent B 1 L dilute Folin reagent

Catalog No. NC9854618 Supplier Bio Rad Laboratories Supplier No. 5000114
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protein Assay Reagent B 1 L dilute Folin reagent

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.