Learn More
Krayden Sylgard 527 Silicone Dielectric Gel | 2lb
Catalog No. NC1208196
Encompass_Preferred
Sylgard 527 Silicone Dielectric Gel is a two component, room temperature and heat curing gel that is used for coating, encapsulating, potting, and sealing electronic devices. It provides flexibility, good flow, and low viscosity.
Catalog No. NC1208196 Supplier Krayden Supplier No. DC1696742
Sylgard 527 Silicone Dielectric Gel is a two component, room temperature and heat curing gel that is used for coating, encapsulating, potting, and sealing electronic devices. It provides flexibility, good flow, and low viscosity.