Learn More
Cayman Chemical DC4 CroslInker 10mg
SDP
Catalog No. 502971162
Encompass_Preferred
A MS-cleavable crosslinking reagent (18Å in length) that contains two intrinsic positive charges, which allow cross-linked peptides to fragment into their component peptides by collision-induced dissociation or in-source decay
Catalog No. 50-297-1162 Supplier Cayman Chemical Supplier No. 1473410 MG
A MS-cleavable crosslinking reagent (18Å in length) that contains two intrinsic positive charges, which allow cross-linked peptides to fragment into their component peptides by collision-induced dissociation or in-source decay