Learn More
Medchemexpress LLC HY-12747 5mg Medchemexpress, DC_517 CAS:500017-70-9 Purity:>98%
SDP
Catalog No. 502081131
Encompass_Preferred
Medchemexpress, HY-12747 5mg DC_517 CAS:500017-70-9 DC_517 is a DNA methyltransferase 1 (DNMT1) inhibitor, with an IC50 and a Kd of 1.7 μM and 0.91 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1131 Supplier Medchemexpress LLC Supplier No. HY127475MG
Medchemexpress, HY-12747 5mg DC_517 CAS:500017-70-9 DC_517 is a DNA methyltransferase 1 (DNMT1) inhibitor, with an IC50 and a Kd of 1.7 μM and 0.91 μM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.