Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Reagent 77 5% w/v Barium Chloride (10L)
SDP

Catalog No. NC9885122
Encompass_Preferred

Reagent 77 5% w/v Barium Chloride (10L)

Catalog No. NC9885122 Supplier Aqua Solutions Supplier No. DC561175
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Reagent 77 5% w/v Barium Chloride (10L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.