Learn More
Selleck Chemical LLC DC661 2mg 1872387-43-3
SDP
Supplier:  Selleck Chemical LLC S88082MG
Encompass_Preferred
DC661 is capable of deacidifying the lysosome and inhibiting autophagy significantly better than hydroxychloroquine (HCQ). DC661 induces apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8145