Learn More
Aviva Systems Biology Corp Dcaf11 Antibody - C-terminal region (ARP42651_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42651_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dcaf11. It was validated on Western Blot. . Alternate names: 0710008A13Rik, C76035, D14Ucla1, Dacf11, GLO14, Wdr23ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-285-015