Learn More
ABclonal Technology DCAF11 Rabbit pAb
SDP
Supplier:  ABclonal Technology A15519500UL
Encompass_Preferred
This gene encodes a WD repeat-containing protein that interacts with the COP9 signalosome, a macromolecular complex that interacts with cullin-RING E3 ligases and regulates their activity by hydrolyzing cullin-Nedd8 conjugates. Multiple alternatively spliced transcript variants have been found for this gene.
Catalog No. 50-001-05991